N-(4-Chloro-2,5-dimethoxyphenyl)-N'-[2-methyl-3-(trifluoromethyl)phenyl]thiourea

Lieferant: Apollo Scientific

PC300918-1G
APOSPC300918-1GEA 452 EUR
APOSPC300918-1G
N-(4-Chloro-2,5-dimethoxyphenyl)-N'-[2-methyl-3-(trifluoromethyl)phenyl]thiourea
N-(4-Chloro-2,5-dimethoxyphenyl)-N'-[2-methyl-3-(trifluoromethyl)phenyl]thiourea
Formel: C₁₇H₁₆ClF₃N₂O₂S
Molecular Weight: 404,834 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30481159
CAS-Nummer: 2198846-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR