3-Chloro-7-(trifluoromethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC300924-1G
APOSPC300924-1GEA 763 EUR
APOSPC300924-1G
3-Chloro-7-(trifluoromethyl)-1H-indazole
3-Chloro-7-(trifluoromethyl)-1H-indazole
Formel: C₈H₄ClF₃N₂
Molecular Weight: 220,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22563698
CAS-Nummer: 1388070-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR