5-Fluoro-3-(4-methoxybenzylidene)-1,3-dihydro-2H-indol-2-one
Lieferant: Apollo Scientific
PC300930-1G
APOSPC300930-1GEA
507
EUR
APOSPC300930-1G
5-Fluoro-3-(4-methoxybenzylidene)-1,3-dihydro-2H-indol-2-one
5-Fluoro-3-(4-methoxybenzylidene)-1,3-dihydro-2H-indol-2-one
Formel:
C₁₆H₁₂FNO₂ Molecular Weight: 269,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14721317 CAS-Nummer: 686726-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...