2-Chloro-5-fluoro-1H-indole-3-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC300936-1G
PC300936-5G
PC300936-250MG
APOSPC300936-1GEA
560
EUR
APOSPC300936-1G
APOSPC300936-5G
APOSPC300936-250MG
2-Chloro-5-fluoro-1H-indole-3-carbaldehyde
2-Chloro-5-fluoro-1H-indole-3-carbaldehyde
Formel:
C₉H₅ClFNO Molecular Weight: 197.594 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05178217 CAS-Nummer: 535925-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...