5-Fluoro-2,6-dimethoxynicotinic acid

Lieferant: Apollo Scientific
Achtung

PC300938-1G
APOSPC300938-1GEA 422 EUR
APOSPC300938-1G
5-Fluoro-2,6-dimethoxynicotinic acid
5-Fluoro-2,6-dimethoxynicotinic acid
Formel: C₈H₈FNO₄
Molecular Weight: 201,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30529705
CAS-Nummer: 2167849-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR