2-Fluoro-1-methoxy-4-(methylsulphonyl)benzene 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC300944-1G
PC300944-250MG
APOSPC300944-1GEA
60.9
EUR
APOSPC300944-1G
APOSPC300944-250MG
2-Fluoro-1-methoxy-4-(methylsulphonyl)benzene 97%
2-Fluoro-1-methoxy-4-(methylsulphonyl)benzene
Formel:
C₈H₉FO₃S Schmelzpunkt: 97…98 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD21090339 CAS-Nummer: 20951-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...