1-[4-(4-Isopropylbenzyl)piperazin-1-yl]-3-(4-fluorophenoxy)propan-1-ol

Lieferant: Apollo Scientific

PC300950-1G
APOSPC300950-1GEA 406 EUR
APOSPC300950-1G
1-[4-(4-Isopropylbenzyl)piperazin-1-yl]-3-(4-fluorophenoxy)propan-1-ol
1-[4-(4-Isopropylbenzyl)piperazin-1-yl]-3-(4-fluorophenoxy)propan-1-ol
Formel: C₂₃H₃₁FN₂O₂
Molecular Weight: 386,503 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29918615
CAS-Nummer: 2109175-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR