4-Chloro-6-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}pyrimidin-2-amine
Lieferant: Apollo Scientific
PC300961-1G
APOSPC300961-1GEA
181
EUR
APOSPC300961-1G
4-Chloro-6-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}pyrimidin-2-amine
4-Chloro-6-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}pyrimidin-2-amine
Formel:
C₁₅H₁₅ClF₃N₅ Molecular Weight: 357,761 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29060040 CAS-Nummer: 2183593-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...