Methyl-2-(4-fluorophenyl)-2-hydroxyacetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(4-fluorophenyl)-2-hydroxyacetate

PC300964-1G
APOSPC300964-1GEA 310 EUR
APOSPC300964-1G
Methyl-2-(4-fluorophenyl)-2-hydroxyacetate
Methyl-2-(4-fluorophenyl)-2-hydroxyacetate
Formel: C₉H₉FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16681137
CAS-Nummer: 127709-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR