6-Bromo-3,4-difluoro-2-nitrophenol

Lieferant: Apollo Scientific
Achtung

PC300970-1G PC300970-250MG
APOSPC300970-1GEA 335 EUR
APOSPC300970-1G APOSPC300970-250MG
6-Bromo-3,4-difluoro-2-nitrophenol
6-Bromo-3,4-difluoro-2-nitrophenol
Formel: C₆H₂BrF₂NO₃
Molecular Weight: 253.99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26099166
CAS-Nummer: 1644281-87-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR