2-(4-Chloro-3-fluorophenoxy)nicotinonitrile
Lieferant: Apollo Scientific
PC300971-1G
APOSPC300971-1GEA
227
EUR
APOSPC300971-1G
2-(4-Chloro-3-fluorophenoxy)nicotinonitrile
2-(4-Chloro-3-fluorophenoxy)nicotinonitrile
Formel:
C₁₂H₆ClFN₂O Molecular Weight: 248,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26178184 CAS-Nummer: 1542060-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...