2-(2-Chloro-4-fluorophenoxy)nicotinonitrile
Lieferant: Apollo Scientific
PC300972-1G
APOSPC300972-1GEA
227
EUR
APOSPC300972-1G
2-(2-Chloro-4-fluorophenoxy)nicotinonitrile
2-(2-Chloro-4-fluorophenoxy)nicotinonitrile
Formel:
C₁₂H₆ClFN₂O Molecular Weight: 248,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13206045 CAS-Nummer: 1183594-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...