N-[2-Methyl-3-(trifluoromethyl)phenyl]-4-(trifluoromethoxy)benzenesulphonamide

Lieferant: Apollo Scientific

PC300974-1G
APOSPC300974-1GEA 373 EUR
APOSPC300974-1G
N-[2-Methyl-3-(trifluoromethyl)phenyl]-4-(trifluoromethoxy)benzenesulphonamide
N-[2-Methyl-3-(trifluoromethyl)phenyl]-4-(trifluoromethoxy)benzenesulphonamide
Formel: C₁₅H₁₁F₆NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD31380627
CAS-Nummer: 2197063-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR