2-(4-Bromo-3-fluorophenoxy)nicotinonitrile

Lieferant: Apollo Scientific

PC300975-1G
APOSPC300975-1GEA 282 EUR
APOSPC300975-1G
2-(4-Bromo-3-fluorophenoxy)nicotinonitrile
2-(4-Bromo-3-fluorophenoxy)nicotinonitrile
Formel: C₁₂H₆BrFN₂O
Molecular Weight: 293,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20975428
CAS-Nummer: 1516062-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR