2-Bromo-4-fluoro-N-methylbenzamide

Lieferant: Apollo Scientific

PC300978-1G
APOSPC300978-1GEA 75.4 EUR
APOSPC300978-1G
2-Bromo-4-fluoro-N-methylbenzamide
2-Bromo-4-fluoro-N-methylbenzamide
Formel: C₈H₇BrFNO
Molecular Weight: 232.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08895950
CAS-Nummer: 923138-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR