2-Bromo-4-fluorobenzohydrazide

Lieferant: Apollo Scientific
Achtung

PC300980-1G
APOSPC300980-1GEA 151 EUR
APOSPC300980-1G
2-Bromo-4-fluorobenzohydrazide
2-Bromo-4-fluorobenzohydrazide
Formel: C₇H₆BrFN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09727562
CAS-Nummer: 935657-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR