4-(Difluoromethoxy)-N,N-dimethylbenzamide

Lieferant: Apollo Scientific

PC300984-1G
APOSPC300984-1GEA 452 EUR
APOSPC300984-1G
4-(Difluoromethoxy)-N,N-dimethylbenzamide
4-(Difluoromethoxy)-N,N-dimethylbenzamide
Formel: C₁₀H₁₁F₂NO₂
Molecular Weight: 215,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20538699
CAS-Nummer: 1378312-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR