2,4-Difluoro-5-iodobenzhydrazide
Lieferant: Apollo Scientific
|
Achtung
|
PC300985-1G
PC300985-250MG
APOSPC300985-1GEA
37.7
EUR
APOSPC300985-1G
APOSPC300985-250MG
2,4-Difluoro-5-iodobenzhydrazide
2,4-Difluoro-5-iodobenzhydrazide
Formel:
C₇H₅F₂IN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630104 CAS-Nummer: 2244085-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...