Ethyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC300986-1G
PC300986-250MG
APOSPC300986-1GEA
130
EUR
APOSPC300986-1G
APOSPC300986-250MG
Ethyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₁H₈BrF₃N₂O₂ Molecular Weight: 337.093 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19704051 CAS-Nummer: 1121051-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...