5-(Trifluoromethoxy)-1H-indole-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

PC300989-1G PC300989-250MG
APOSPC300989-1GEA 1880 EUR
APOSPC300989-1G APOSPC300989-250MG
5-(Trifluoromethoxy)-1H-indole-2-carbohydrazide
5-(Trifluoromethoxy)-1H-indole-2-carbohydrazide
Formel: C₁₀H₈F₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04110840
CAS-Nummer: 685120-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR