2,2-Dimethyl-N-[2-(trifluoromethoxy)phenyl]propanamide

Lieferant: Apollo Scientific

PC300990-1G
APOSPC300990-1GEA 227 EUR
APOSPC300990-1G
2,2-Dimethyl-N-[2-(trifluoromethoxy)phenyl]propanamide
2,2-Dimethyl-N-[2-(trifluoromethoxy)phenyl]propanamide
Formel: C₁₂H₁₄F₃NO₂
Molecular Weight: 261.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05975031
CAS-Nummer: 860784-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR