3-(Dimethylamino)-1-(2-fluoro-1,1'-biphenyl-4-yl)prop-2-en-1-one

Lieferant: Apollo Scientific

PC300992-1G
APOSPC300992-1GEA 226 EUR
APOSPC300992-1G
3-(Dimethylamino)-1-(2-fluoro-1,1'-biphenyl-4-yl)prop-2-en-1-one
3-(Dimethylamino)-1-(2-fluoro-1,1'-biphenyl-4-yl)prop-2-en-1-one
Formel: C₁₇H₁₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD18089276
CAS-Nummer: 1287227-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR