3-(2-Fluoro-1,1'-biphenyl-4-yl)-1H-pyrazole

Lieferant: Apollo Scientific

PC300993-1G
APOSPC300993-1GEA 226 EUR
APOSPC300993-1G
3-(2-Fluoro-1,1'-biphenyl-4-yl)-1H-pyrazole
3-(2-Fluoro-1,1'-biphenyl-4-yl)-1H-pyrazole
Formel: C₁₅H₁₁FN₂
Molecular Weight: 238,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18089275
CAS-Nummer: 1287217-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR