4-Chloro-N-(3-fluoro-4-methylphenyl)butanamide
Lieferant: Apollo Scientific
PC300995-1G
APOSPC300995-1GEA
227
EUR
APOSPC300995-1G
4-Chloro-N-(3-fluoro-4-methylphenyl)butanamide
4-Chloro-N-(3-fluoro-4-methylphenyl)butanamide
Formel:
C₁₁H₁₃ClFNO Molecular Weight: 229,678 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09730949 CAS-Nummer: 953746-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...