5-Chloro-2-fluoro-N-methylbenzamide

Lieferant: Apollo Scientific
Achtung

PC300996-100MG
APOSPC300996-100MGEA 21.9 EUR
APOSPC300996-100MG
5-Chloro-2-fluoro-N-methylbenzamide
5-Chloro-2-fluoro-N-methylbenzamide
Formel: C₈H₇ClFNO
Molecular Weight: 187,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14587408
CAS-Nummer: 1223412-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR