N-{[4-Bromo-2-(trifluoromethoxy)phenyl]carbamothioyl}benzamide
Lieferant: Apollo Scientific
PC300999-1G
APOSPC300999-1GEA
452
EUR
APOSPC300999-1G
N-{[4-Bromo-2-(trifluoromethoxy)phenyl]carbamothioyl}benzamide
N-{[4-Bromo-2-(trifluoromethoxy)phenyl]carbamothioyl}benzamide
Formel:
C₁₅H₁₀BrF₃N₂O₂S Molecular Weight: 419,216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630106 CAS-Nummer: 2244085-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...