Ethyl 2-(trifluoromethyl)imidazo[1,2-a]pyrimidine-3-carboxylate

Lieferant: Apollo Scientific

PC301012-1G
APOSPC301012-1GEA 564 EUR
APOSPC301012-1G
Ethyl 2-(trifluoromethyl)imidazo[1,2-a]pyrimidine-3-carboxylate
Ethyl 2-(trifluoromethyl)imidazo[1,2-a]pyrimidine-3-carboxylate
Formel: C₁₀H₈F₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05668962
CAS-Nummer: 866149-79-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR