2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC301013-1G
APOSPC301013-1GEA 373 EUR
APOSPC301013-1G
2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine-3-carboxylic acid
2-(Trifluoromethyl)imidazo[1,2-a]pyrimidine-3-carboxylic acid
Formel: C₈H₄F₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05668980
CAS-Nummer: 866149-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR