1-(4-Fluorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone

Lieferant: Apollo Scientific

PC301016-1G
APOSPC301016-1GEA 493 EUR
APOSPC301016-1G
1-(4-Fluorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
1-(4-Fluorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
Formel: C₁₄H₁₀Cl₃FN₂
Molecular Weight: 331,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01132766
CAS-Nummer: 394695-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR