Ethyl (4-bromo-2-fluoro-5-methylphenoxy)acetate
Lieferant: Apollo Scientific
PC301018-1G
APOSPC301018-1GEA
310
EUR
APOSPC301018-1G
Ethyl (4-bromo-2-fluoro-5-methylphenoxy)acetate
Ethyl (4-bromo-2-fluoro-5-methylphenoxy)acetate
Formel:
C₁₁H₁₂BrFO₃ Molecular Weight: 291,114 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31725299 CAS-Nummer: 2270912-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...