(4-Bromo-2-fluoro-5-methylphenoxy)acetic acid

Lieferant: Apollo Scientific

PC301019-1G
APOSPC301019-1GEA 371 EUR
APOSPC301019-1G
(4-Bromo-2-fluoro-5-methylphenoxy)acetic acid
(4-Bromo-2-fluoro-5-methylphenoxy)acetic acid
Formel: C₉H₈BrFO₃
Molecular Weight: 263,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31725300
CAS-Nummer: 2270912-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR