1-(Benzyloxy)-4-bromo-2-fluoro-5-methylbenzene

Lieferant: Apollo Scientific

PC301021-1G
APOSPC301021-1GEA 371 EUR
APOSPC301021-1G
1-(Benzyloxy)-4-bromo-2-fluoro-5-methylbenzene
1-(Benzyloxy)-4-bromo-2-fluoro-5-methylbenzene
Formel: C₁₄H₁₂BrFO
Molecular Weight: 295,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31725302
CAS-Nummer: 2234291-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR