Ethyl (2-fluoro-5-methylphenoxy)acetate
Lieferant: Apollo Scientific
PC301022-1G
APOSPC301022-1GEA
223
EUR
APOSPC301022-1G
Ethyl (2-fluoro-5-methylphenoxy)acetate
Ethyl (2-fluoro-5-methylphenoxy)acetate
Formel:
C₁₁H₁₃FO₃ Molecular Weight: 212,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18911516 CAS-Nummer: 1339681-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...