(2-Fluoro-5-methylphenoxy)acetic acid
Lieferant: Apollo Scientific
PC301023-1G
APOSPC301023-1GEA
248
EUR
APOSPC301023-1G
(2-Fluoro-5-methylphenoxy)acetic acid
(2-Fluoro-5-methylphenoxy)acetic acid
Formel:
C₉H₉FO₃ Molecular Weight: 184,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20429669 CAS-Nummer: 1394793-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...