Ethyl 8-bromo-2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate

Lieferant: Apollo Scientific

PC301038-1G PC301038-250MG
APOSPC301038-1GEA 522 EUR
APOSPC301038-1G APOSPC301038-250MG
Ethyl 8-bromo-2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 8-bromo-2-(difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₂H₁₁BrF₂N₂O₂
Schmelzpunkt: 128…131 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2451256-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR