Ethyl 8-bromo-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Lieferant: Apollo Scientific

PC301042-1G PC301042-250MG
APOSPC301042-1GEA 522 EUR
APOSPC301042-1G APOSPC301042-250MG
Ethyl 8-bromo-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 8-bromo-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₁H₉BrF₂N₂O₂
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2451256-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR