Dimethyl dodecafluorosuberate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dimethyl perfluorosuberate
PC3011-100G
PC3011-25G
PC3011-5G
APOSPC3011-100GEA
1360
EUR
APOSPC3011-100G
APOSPC3011-25G
APOSPC3011-5G
Dimethyl dodecafluorosuberate
Dimethyl dodecafluorosuberate
Formel:
C₁₀H₆F₁₂O₄ Siedepunkt: 152 °C/60mmHg Schmelzpunkt: 26 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044088 CAS-Nummer: 2062-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...