Dimethyl perfluoro-1,10-decanedicarboxylate 96%

Lieferant: Apollo Scientific

PC3016H-5G
APOSPC3016H-5GEA 116 EUR
APOSPC3016H-5G
Dimethyl perfluoro-1,10-decanedicarboxylate 96%
Dimethyl perfluoro-1,10-decanedicarboxylate
Formel: C₁₄H₆F₂₀O₄
Molecular Weight: 618,17 g/mol
Schmelzpunkt: 31…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236606
CAS-Nummer: 84750-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR