5-Fluor-2-(trifluormethyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

PC3019-500MG
APOSPC3019-500MGEA 0 EUR
APOSPC3019-500MG
5-Fluor-2-(trifluormethyl)phenylboronsäure
5-Fluor-2-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BF₄O₂
Lagertemperatur: Kühlschrank
MDL: MFCD06656275
CAS-Nummer: 928053-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR