2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol 98%

Lieferant: Apollo Scientific

PC302018-1G PC302018-5G
APOSPC302018-1GEA 0 EUR
APOSPC302018-1G APOSPC302018-5G
2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol 98%
2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol
Formel: C₉H₈F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD06660357
CAS-Nummer: 886761-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR