2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol 98%

Lieferant: Apollo Scientific

PC302019-1G PC302019-5G PC302019-25G PC302019-100G
APOSPC302019-1GEA 0 EUR
APOSPC302019-1G APOSPC302019-5G APOSPC302019-25G APOSPC302019-100G
2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol 98%
2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol
Formel: C₉H₈F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD06660358
CAS-Nummer: 886761-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR