2-Chloro-4,5-difluorobenzamide 98%

Lieferant: Apollo Scientific

PC302021-5G PC302021-1G
APOSPC302021-5GEA 255 EUR
APOSPC302021-5G APOSPC302021-1G
2-Chloro-4,5-difluorobenzamide 98%
2-Chloro-4,5-difluorobenzamide
Formel: C₇H₄ClF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD02628151
CAS-Nummer: 296274-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR