4-Ethoxy-2,6-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC302024-1G
APOSPC302024-1GEA 1630 EUR
APOSPC302024-1G
4-Ethoxy-2,6-difluorobenzenesulphonyl chloride
4-Ethoxy-2,6-difluorobenzenesulphonyl chloride
Formel: C₈H₇ClF₂O₃S
Molecular Weight: 256,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054226
CAS-Nummer: 1432129-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR