2-Chloro-6-(trifluoromethyl)phenylacetonitrile 97%

Lieferant: Apollo Scientific
Gefahr

PC302025-1G PC302025-5G PC302025-25G PC302025-100G
APOSPC302025-1GEA 0 EUR
APOSPC302025-1G APOSPC302025-5G APOSPC302025-25G APOSPC302025-100G
2-Chloro-6-(trifluoromethyl)phenylacetonitrile 97%
Formel: C₉H₅ClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD06660233
CAS-Nummer: 633317-69-8
UN: 3439
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR