2-Fluoro-5-(trifluoromethoxy)cinnamic acid

Lieferant: Apollo Scientific

PC302026-5G
APOSPC302026-5GEA 372 EUR
APOSPC302026-5G
2-Fluoro-5-(trifluoromethoxy)cinnamic acid
2-Fluoro-5-(trifluoromethoxy)cinnamic acid
Formel: C₁₀H₆F₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09832239
CAS-Nummer: 1017777-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR