3-(3-Chloro-5-fluorophenyl)propionic acid 97%

Lieferant: Apollo Scientific

PC302033-1G
APOSPC302033-1GEA 0 EUR
APOSPC302033-1G
3-(3-Chloro-5-fluorophenyl)propionic acid 97%
3-(3-Chloro-5-fluorophenyl)propionic acid
Formel: C₉H₈ClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04116057
CAS-Nummer: 886498-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR