3,4-Difluorbenzamidoxim

Lieferant: Apollo Scientific

PC302036-10G PC302036-50G
APOSPC302036-10GEA 79.9 EUR
APOSPC302036-10G APOSPC302036-50G
3,4-Difluorbenzamidoxim
3,4-Difluorbenzamidoxim
Formel: C₇H₆F₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD06660194
CAS-Nummer: 500024-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR