3-Chlor-4-(trifluormethyl)zimtsäure 97%

Lieferant: Apollo Scientific

PC302047-1G
APOSPC302047-1GEA 0 EUR
APOSPC302047-1G
3-Chlor-4-(trifluormethyl)zimtsäure 97%
3-Chlor-4-(trifluormethyl)zimtsäure
Formel: C₁₀H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11519328
CAS-Nummer: 1092460-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR