3-Chloro-5-(trifluoromethoxy)cinnamic acid 97%
Lieferant: Apollo Scientific
PC302049-1G
APOSPC302049-1GEA
0
EUR
APOSPC302049-1G
3-Chloro-5-(trifluoromethoxy)cinnamic acid 97%
3-Chloro-5-(trifluoromethoxy)cinnamic acid
Formel:
C₁₀H₆ClF₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660339 CAS-Nummer: 1588505-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...