3-Chloro-5-(trifluoromethyl)cinnamic acid 95%

Lieferant: Apollo Scientific

PC302051-1G PC302051-5G PC302051-25G
APOSPC302051-1GEA 0 EUR
APOSPC302051-1G APOSPC302051-5G APOSPC302051-25G
3-Chloro-5-(trifluoromethyl)cinnamic acid 95%
3-Chloro-5-(trifluoromethyl)cinnamic acid
Formel: C₁₀H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660340
CAS-Nummer: 886761-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR